siRNA Details
Gene Symbol: Tyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106664.1 | NP_001100134.1 | 2 | 309 | ||
XM_006238332.2 | XP_006238394.1 | 2 | 340 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106664.1 | NP_001100134.1 | 2 | 309 | ||
XM_006238332.2 | XP_006238394.1 | 2 | 340 |