siRNA Details
Gene Symbol: Plin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007144.1 | NP_001007145.1 | 6, 7 | 843 | ||
XM_006238360.2 | XP_006238422.1 | 6, 7 | 879 | |||
XM_006238362.2 | XP_006238424.1 | 5, 6 | 701 | |||
GenBank mRNA | AW913874.1 | - | 1, 2 | 160 | ||
BC085861.1 | - | 6, 7 | 843 | 7190361 |
