siRNA Details
Gene Symbol: Yipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199383.2 | NP_955415.1 | 3 | 196 | ||
XM_006238504.2 | XP_006238566.1 | 3 | 234 | |||
XM_006238505.2 | XP_006238567.1 | 3 | 233 | |||
GenBank mRNA | BC062239.1 | - | 3 | 200 | 6887537 | |
FQ218908.1 | - | 3 | 195 |
