siRNA Details
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004236.1 | NP_001004236.1 | 6 | 676 | ||
XM_006238639.1 | XP_006238701.1 | 8 | 949 | |||
GenBank mRNA | BC078938.1 | - | 6 | 676 | 7108356 |
