siRNA Details
Gene Symbol: Toe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106680.1 | NP_001100150.1 | 6 | 725 | ||
XM_006238651.1 | XP_006238713.1 | 6 | 720 | |||
XM_006238652.2 | XP_006238714.1 | 5 | 1050 | |||
XM_006238653.2 | XP_006238715.1 | 5 | 972 | |||
XM_006238654.2 | XP_006238716.1 | 5 | 1073 |
