siRNA Details
Gene Symbol: Eif4g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106693.1 | NP_001100163.1 | 9 | 946 | ||
XM_008764221.1 | XP_008762443.1 | 12 | 1294 | |||
XM_008764222.1 | XP_008762444.1 | 10 | 1243 | |||
XM_008764223.1 | XP_008762445.1 | 11 | 1183 | |||
XM_008764224.1 | XP_008762446.1 | 10 | 1162 | |||
XM_008764225.1 | XP_008762447.1 | 10 | 1147 | |||
XM_008764226.1 | XP_008762448.1 | 9 | 1128 | |||
XM_008764227.1 | XP_008762449.1 | 9 | 1114 | |||
XM_008764228.1 | XP_008762450.1 | 8 | 1032 | |||
XM_008764229.1 | XP_008762451.1 | 12 | 1123 |
