siRNA Details
Gene Symbol: Eif4g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106693.1 | NP_001100163.1 | 15 | 2632 | ||
XM_008764221.1 | XP_008762443.1 | 18 | 2980 | |||
XM_008764222.1 | XP_008762444.1 | 16 | 2929 | |||
XM_008764223.1 | XP_008762445.1 | 17 | 2869 | |||
XM_008764224.1 | XP_008762446.1 | 16 | 2848 | |||
XM_008764225.1 | XP_008762447.1 | 16 | 2833 | |||
XM_008764226.1 | XP_008762448.1 | 15 | 2814 | |||
XM_008764227.1 | XP_008762449.1 | 15 | 2800 | |||
XM_008764228.1 | XP_008762450.1 | 14 | 2718 | |||
XM_008764229.1 | XP_008762451.1 | 18 | 2809 |
