siRNA Details
Gene Symbol: Tmem82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239327.2 | XP_006239389.1 | 5 | 1544 | ||
XM_008764305.1 | XP_008762527.1 | 5 | 1685 | |||
XM_008764306.1 | XP_008762528.1 | 5 | 1685 | |||
XM_238429.8 | XP_238429.5 | 5 | 1544 |
