siRNA Details
Gene Symbol: Ccnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013094.1 | NP_001013112.1 | 1 | 133 | ||
XM_006239544.2 | XP_006239606.1 | 1 | 655 | |||
XM_006239545.2 | XP_006239607.1 | 1 | 654 | |||
GenBank mRNA | BC088316.1 | - | 1 | 133 | 7325177 | |
FQ211043.1 | - | 1 | 148 |
