siRNA Details
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013095.1 | NP_001013113.1 | 4 | 205 | ||
GenBank mRNA | BC086383.1 | - | 4 | 205 | 7301572 | |
FQ213005.1 | - | 4 | 246 | |||
FQ225883.1 | - | 4 | 238 | |||
FQ226276.1 | - | 4 | 237 | |||
FQ229703.1 | - | 4 | 236 | |||
FQ230888.1 | - | 4 | 231 | |||
FQ232595.1 | - | 4 | 235 | |||
FQ232661.1 | - | 4 | 235 | |||
FQ234727.1 | - | 2 | 87 | |||
FQ234921.1 | - | 4 | 236 | |||
FQ234929.1 | - | 4 | 238 | |||
FQ235264.1 | - | 1 | 19 |
