siRNA Details
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010951.1 | NP_001010951.1 | 9 | 1313 | ||
XM_006239697.2 | XP_006239759.1 | 10 | 1876 | |||
XM_006239698.2 | XP_006239760.2 | 10 | 1441 | |||
XM_006239699.2 | XP_006239761.2 | 10 | 2153 | |||
XM_008764456.1 | XP_008762678.1 | 10 | 1543 | |||
XM_008764457.1 | XP_008762679.1 | 10 | 1436 | |||
XM_008764458.1 | XP_008762680.1 | 9 | 1236 | |||
GenBank mRNA | BC089111.1 | - | 9 | 1313 | 7312962 | |
FQ213992.1 | - | 3 | 352 |
