siRNA Details
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106710.1 | NP_001100180.1 | 4 | 2278 | ||
XM_006239793.2 | XP_006239855.1 | 4 | 2289 | |||
XM_008764511.1 | XP_008762733.1 | 4 | 3021 |

More Information
Other Species Targeted

Gene Ontology Categories:
