siRNA Details
Gene Symbol: Mapre3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007656.1 | NP_001007657.1 | 5 | 536 | ||
XM_008764512.1 | XP_008762734.1 | 4 | 586 | |||
GenBank mRNA | BC083589.1 | - | 5 | 536 | 7132546 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133350, AF468005, U51204, BC057918.
Gene Ontology Categories:
