siRNA Details
Gene Symbol: Iah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100540.1 | NP_001094010.1 | 4, 5 | 452 | ||
GenBank mRNA | BC168771.1 | - | 4, 5 | 452 | 6919650 | |
FQ220941.1 | - | 4, 5 | 448 | |||
FQ225724.1 | - | 4, 5 | 477 | |||
FQ231316.1 | - | 4, 5 | 477 | |||
FQ232084.1 | - | 4, 5 | 482 | |||
FQ235324.1 | - | 4, 5 | 477 |
