siRNA Details
Gene Symbol: Iah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100540.1 | NP_001094010.1 | 3 | 257 | ||
GenBank mRNA | AJ344542.1 | - | 2 | 182 | ||
BC168771.1 | - | 3 | 257 | 6919650 | ||
FQ220941.1 | - | 3 | 253 | |||
FQ231316.1 | - | 3 | 282 | |||
FQ232084.1 | - | 3 | 287 | |||
FQ235324.1 | - | 3 | 282 |
