siRNA Details
Gene Symbol: Scin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198748.1 | NP_942043.1 | 5 | 722 | ||
XM_008764644.1 | XP_008762866.1 | 5 | 1060 | |||
XM_008764645.1 | XP_008762867.1 | 3 | 535 | |||
GenBank mRNA | AY324394.1 | - | 5 | 722 |
