siRNA Details
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185025.1 | NP_001171954.1 | 6 | 474 | ||
XM_008764655.1 | XP_008762877.1 | 6 | 429 | |||
GenBank mRNA | CO399813.1 | - | 6 | 450 | ||
FQ210674.1 | - | 6 | 492 |
