siRNA Details
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185025.1 | NP_001171954.1 | 11 | 929 | ||
XM_008764655.1 | XP_008762877.1 | 11 | 884 | |||
GenBank mRNA | BF282469.1 | - | 1 | 225 | ||
FQ210674.1 | - | 11 | 947 |
