siRNA Details
Gene Symbol: Brms1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106731.1 | NP_001100201.1 | 467 | |||
GenBank mRNA | BC162034.1 | - | 478 | 7096422 |
Gene Symbol: LOC100910898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750158.2 | XP_003750206.1 | 3, 4 | 497 |
