siRNA Details
Gene Symbol: Fkbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106736.2 | NP_001100206.2 | 3, 4 | 341 | ||
GenBank mRNA | BC168873.1 | - | 3, 4 | 306 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fkbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106736.2 | NP_001100206.2 | 3, 4 | 341 | ||
GenBank mRNA | BC168873.1 | - | 3, 4 | 306 |