siRNA Details
Gene Symbol: Pole2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169108.1 | NP_001162579.1 | 17 | 1459 | ||
XM_006240154.2 | XP_006240216.1 | 17 | 1471 | |||
XM_008764696.1 | XP_008762918.1 | 20 | 2258 | |||
XM_008764697.1 | XP_008762919.1 | 19 | 2068 | |||
XM_008764698.1 | XP_008762920.1 | 20 | 2197 | |||
XM_008764699.1 | XP_008762921.1 | 20 | 2281 | |||
XM_008764700.1 | XP_008762922.1 | 20 | 2167 | |||
XM_008764701.1 | XP_008762923.1 | 20 | 2061 | |||
XM_008764702.1 | XP_008762924.1 | 20 | 2157 | |||
GenBank mRNA | FQ231562.1 | - | 11 | 895 |
