siRNA Details
Gene Symbol: Nemf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170586.1 | NP_001164057.1 | 5 | 536 | ||
GenBank mRNA | CA512507.1 | - | 4 | 375 | ||
CB545170.1 | - | 2 | 194 | |||
FM071452.1 | - | 1 | 52 | |||
FQ205447.1 | - | 2 | 156 |
