siRNA Details
Gene Symbol: Nin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106737.1 | NP_001100207.1 | 14 | 1786 | ||
XM_006240158.2 | XP_006240220.1 | 15 | 1946 | |||
XM_008764703.1 | XP_008762925.1 | 14 | 2079 | |||
XM_008764704.1 | XP_008762926.1 | 15 | 1946 | |||
XM_008764705.1 | XP_008762927.1 | 14 | 1856 | |||
XM_008764706.1 | XP_008762928.1 | 14 | 1856 | |||
XM_008764707.1 | XP_008762929.1 | 15 | 1946 | |||
XM_008764708.1 | XP_008762930.1 | 14 | 1856 | |||
XM_008764709.1 | XP_008762931.1 | 15 | 1946 | |||
XM_008764710.1 | XP_008762932.1 | 14 | 1854 |
Gene Symbol: LOC100911256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240147.2 | XP_006240209.1 | 1438 |
