siRNA Details
Gene Symbol: LOC100911256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240147.2 | XP_006240209.1 | 3565 |
Gene Symbol: Nin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106737.1 | NP_001100207.1 | 18 | 3913 | ||
XM_006240158.2 | XP_006240220.1 | 19 | 4073 | |||
XM_008764703.1 | XP_008762925.1 | 18 | 4206 | |||
XM_008764704.1 | XP_008762926.1 | 19 | 4073 | |||
XM_008764705.1 | XP_008762927.1 | 18 | 3983 | |||
XM_008764706.1 | XP_008762928.1 | 18 | 3983 | |||
XM_008764707.1 | XP_008762929.1 | 19 | 4073 | |||
XM_008764708.1 | XP_008762930.1 | 18 | 3983 | |||
XM_008764709.1 | XP_008762931.1 | 19 | 4073 |
