siRNA Details
Gene Symbol: Actr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009602.1 | NP_001009602.1 | 5, 6 | 468 | ||
GenBank mRNA | BC087143.1 | - | 5, 6 | 468 | 7128051 | |
FQ219976.1 | - | 1, 2 | 106 | |||
FQ228754.1 | - | 5, 6 | 523 |
