siRNA Details
Gene Symbol: Dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191818.1 | NP_001178747.1 | 8 | 784 | ||
XM_008764746.1 | XP_008762968.1 | 9 | 1004 | |||
XM_008764747.1 | XP_008762969.1 | 10 | 985 | |||
XM_008764748.1 | XP_008762970.1 | 9 | 1003 | |||
XM_008764749.1 | XP_008762971.1 | 8 | 833 | |||
XM_008764750.1 | XP_008762972.1 | 9 | 1003 | |||
XM_008764751.1 | XP_008762973.1 | 8 | 1250 |
