siRNA Details
Gene Symbol: Arel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106744.1 | NP_001100214.1 | 15 | 2142 | ||
XM_006240333.2 | XP_006240395.1 | 16 | 3128 | |||
XM_006240335.2 | XP_006240397.1 | 16 | 3098 | |||
XM_008764754.1 | XP_008762976.1 | 15 | 2403 | |||
XM_008764755.1 | XP_008762977.1 | 17 | 3235 | |||
GenBank mRNA | FQ226603.1 | - | 7 | 721 |
