siRNA Details
Gene Symbol: Ttc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106752.1 | NP_001100222.1 | 11 | 1341 | ||
XM_006240422.2 | XP_006240484.1 | 13 | 1429 | |||
XM_006240423.2 | XP_006240485.1 | 12 | 1398 | |||
XM_006240424.2 | XP_006240486.1 | 13 | 1407 | |||
XM_008764762.1 | XP_008762984.1 | 11 | 1187 | |||
GenBank mRNA | BC161880.1 | - | 12 | 1373 | 7126409 |
