siRNA Details
Gene Symbol: Ttc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106752.1 | NP_001100222.1 | 9 | 972 | ||
XM_006240422.2 | XP_006240484.1 | 11 | 1060 | |||
XM_006240423.2 | XP_006240485.1 | 10 | 1029 | |||
XM_006240424.2 | XP_006240486.1 | 11 | 1038 | |||
XM_008764762.1 | XP_008762984.1 | 9 | 818 | |||
GenBank mRNA | BC161880.1 | - | 10 | 1004 | 7126409 |
