siRNA Details
Gene Symbol: Serpina6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009663.1 | NP_001009663.1 | 5 | 1238 | ||
GenBank mRNA | BC088300.1 | - | 5 | 1238 | 7323342 | |
FQ209415.1 | - | 5 | 1283 | |||
FQ211289.1 | - | 5 | 1253 | |||
FQ218294.1 | - | 5 | 1261 | |||
FQ218398.1 | - | 5 | 1248 | |||
FQ219289.1 | - | 5 | 1313 |
