siRNA Details
Gene Symbol: Akap8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013946.1 | NP_001013968.1 | 4, 5 | 443 | ||
XM_006241054.1 | XP_006241116.1 | 5, 6 | 351 | |||
XM_006241055.1 | XP_006241117.1 | 4, 5 | 453 | |||
GenBank mRNA | BC085785.1 | - | 4, 5 | 443 | 7108413 |
