siRNA Details
Gene Symbol: Casp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191776.1 | NP_001178705.1 | 2 | 138 | ||
XM_006241046.2 | XP_006241108.1 | 3 | 336 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Casp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191776.1 | NP_001178705.1 | 2 | 138 | ||
XM_006241046.2 | XP_006241108.1 | 3 | 336 |