siRNA Details
Gene Symbol: Polrmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106766.1 | NP_001100236.1 | 7 | 1373 | ||
XM_006240859.2 | XP_006240921.1 | 7 | 1402 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Polrmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106766.1 | NP_001100236.1 | 7 | 1373 | ||
XM_006240859.2 | XP_006240921.1 | 7 | 1402 |