siRNA Details
Gene Symbol: Abca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207598.1 | NP_997481.1 | 36 | 5130 | ||
XM_006240871.2 | XP_006240933.1 | 37 | 5484 | |||
XM_006240872.2 | XP_006240934.1 | 37 | 5381 | |||
XM_006240873.2 | XP_006240935.1 | 37 | 5372 | |||
XM_006240874.2 | XP_006240936.1 | 37 | 5269 | |||
XM_006240875.2 | XP_006240937.1 | 35 | 5223 | |||
GenBank mRNA | AB097814.1 | - | 36 | 5130 |
