siRNA Details
Gene Symbol: Mknk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011985.1 | NP_001011985.1 | 2 | 164 | ||
XM_006240883.2 | XP_006240945.1 | 2 | 253 | |||
XM_006240884.1 | XP_006240946.1 | 1 | 79 | |||
GenBank mRNA | BC085941.1 | - | 2 | 164 | 7124839 | |
FQ211808.1 | - | 2 | 207 |
