siRNA Details
Gene Symbol: Lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765148.1 | XP_008763370.1 | 4 | 1255 | ||
XM_008765149.1 | XP_008763371.1 | 4 | 1526 | |||
XM_216850.8 | XP_216850.6 | 3 | 1667 | |||
GenBank mRNA | FQ212316.1 | - | 3 | 491 |
