siRNA Details
Gene Symbol: Tle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039013.1 | NP_001034102.1 | 19 | 2109 | ||
XM_006240889.2 | XP_006240951.1 | 20 | 2422 | |||
XM_006240890.2 | XP_006240952.1 | 20 | 2899 | |||
XM_006240892.2 | XP_006240954.1 | 20 | 2335 | |||
XM_006240893.2 | XP_006240955.1 | 20 | 2419 | |||
XM_006240894.2 | XP_006240956.1 | 20 | 2386 | |||
XM_006240895.2 | XP_006240957.1 | 20 | 2491 | |||
GenBank mRNA | BC100652.1 | - | 19 | 2109 | 7932424 |
