siRNA Details
Gene Symbol: Nsmce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024876.1 | NP_001020047.1 | 3 | 302 | ||
XM_006241671.2 | XP_006241733.1 | 3 | 342 | |||
XM_008765505.1 | XP_008763727.1 | 2 | 2462 | |||
XM_008765506.1 | XP_008763728.1 | 3 | 342 | |||
GenBank mRNA | BC097477.1 | - | 3 | 302 | 7458086 |
