siRNA Details
Gene Symbol: Nsmce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024876.1 | NP_001020047.1 | 5, 6 | 597 | ||
XM_008765505.1 | XP_008763727.1 | 4, 5 | 2757 | |||
GenBank mRNA | BC097477.1 | - | 5, 6 | 597 | 7458086 |
