siRNA Details
Gene Symbol: Tsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127455.2 | NP_001120927.1 | 3 | 248 | ||
NM_001270789.1 | NP_001257718.1 | 3 | 314 | |||
NM_001270790.1 | NP_001257719.1 | 3 | 314 | |||
GenBank mRNA | BC158885.1 | - | 3 | 286 | 8363456 | |
BF289545.1 | - | 3 | 249 | |||
CA339313.1 | - | 2 | 202 | |||
CK653514.1 | - | 247 | ||||
FQ210097.1 | - | 3 | 321 | |||
FQ221078.1 | - | 3 | 236 | |||
FQ229919.1 | - | 3 | 259 |
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ210517.1 | - | 3862 |
