siRNA Details
Gene Symbol: Tsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127455.2 | NP_001120927.1 | 10 | 920 | ||
NM_001270789.1 | NP_001257718.1 | 10 | 986 | |||
NM_001270790.1 | NP_001257719.1 | 9 | 857 | |||
GenBank mRNA | BC158885.1 | - | 9 | 829 | 8363456 | |
FQ210097.1 | - | 10 | 993 | |||
FQ221078.1 | - | 10 | 909 | |||
FQ229919.1 | - | 10 | 933 |
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ210517.1 | - | 3190 |
