siRNA Details
Gene Symbol: Tsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127455.2 | NP_001120927.1 | 2 | 69 | ||
NM_001270789.1 | NP_001257718.1 | 2 | 135 | |||
NM_001270790.1 | NP_001257719.1 | 2 | 135 | |||
GenBank mRNA | BC158885.1 | - | 2 | 107 | 8363456 | |
BF289545.1 | - | 2 | 70 | |||
CA339313.1 | - | 1 | 23 | |||
CK653514.1 | - | 68 | ||||
FQ210097.1 | - | 2 | 142 | |||
FQ221078.1 | - | 2 | 57 | |||
FQ229919.1 | - | 2 | 80 |
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ210517.1 | - | 4041 |
