siRNA Details
Gene Symbol: Recql4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130494.1 | NP_001123966.1 | 14 | 2201 | ||
XM_006241806.2 | XP_006241868.1 | 14 | 2276 | |||
XM_008765566.1 | XP_008763788.1 | 14 | 2294 | |||
XM_008765567.1 | XP_008763789.1 | 14 | 2291 | |||
XM_008765568.1 | XP_008763790.1 | 14 | 2294 |
