siRNA Details
Gene Symbol: Ppp6r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106789.1 | NP_001100259.1 | 13 | 1831 | ||
XM_008765688.1 | XP_008763910.1 | 9 | 1152 | |||
GenBank mRNA | BC166525.1 | - | 9 | 1289 | ||
FQ211682.1 | - | 1538 | ||||
FQ230825.1 | - | 12 | 1750 |
