siRNA Details
Gene Symbol: Ddx23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106793.2 | NP_001100263.2 | 7 | 791 | ||
XM_006257343.2 | XP_006257405.1 | 7 | 880 | |||
GenBank mRNA | BC169082.1 | - | 6 | 670 |
