siRNA Details
Gene Symbol: Aasdhppt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106798.2 | NP_001100268.2 | 4, 5 | 708 | ||
GenBank mRNA | BC098729.1 | - | 3, 4 | 1064 | ||
BC166800.1 | - | 4, 5 | 708 | 7938259 | ||
FQ209549.1 | - | 4, 5 | 724 | |||
FQ214028.1 | - | 4, 5 | 727 | |||
FQ214274.1 | - | 4, 5 | 721 | |||
FQ230080.1 | - | 4, 5 | 727 |
