siRNA Details
Gene Symbol: Naalad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106802.1 | NP_001100272.1 | 22 | 2454 | ||
XM_006242562.2 | XP_006242624.1 | 22 | 2802 | |||
XM_006242568.2 | XP_006242630.1 | 21 | 2668 | |||
XM_008765922.1 | XP_008764144.1 | 19 | 2311 | |||
XM_008765923.1 | XP_008764145.1 | 18 | 2212 | |||
XM_008765924.1 | XP_008764146.1 | 20 | 2422 | |||
XM_008765925.1 | XP_008764147.1 | 22 | 2319 | |||
XM_008765926.1 | XP_008764148.1 | 21 | 2655 | |||
XM_008765927.1 | XP_008764149.1 | 13 | 1260 | |||
GenBank mRNA | BC168965.1 | - | 20 | 2228 |

More Information
Other Species Targeted

Gene Ontology Categories:
