siRNA Details
Gene Symbol: Naalad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106802.1 | NP_001100272.1 | 11 | 1292 | ||
XM_006242562.2 | XP_006242624.1 | 11 | 1640 | |||
XM_006242568.2 | XP_006242630.1 | 10 | 1506 | |||
XM_008765922.1 | XP_008764144.1 | 8 | 1149 | |||
XM_008765924.1 | XP_008764146.1 | 9 | 1260 | |||
XM_008765925.1 | XP_008764147.1 | 11 | 1157 | |||
XM_008765926.1 | XP_008764148.1 | 10 | 1493 | |||
XM_008765927.1 | XP_008764149.1 | 2 | 98 | |||
GenBank mRNA | BC168965.1 | - | 9 | 1070 |

More Information
Other Species Targeted

Gene Ontology Categories:
