siRNA Details
Gene Symbol: Naalad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106802.1 | NP_001100272.1 | 9 | 1047 | ||
XM_006242562.2 | XP_006242624.1 | 9 | 1395 | |||
XM_006242568.2 | XP_006242630.1 | 8 | 1261 | |||
XM_008765922.1 | XP_008764144.1 | 6 | 904 | |||
XM_008765923.1 | XP_008764145.1 | 6 | 904 | |||
XM_008765924.1 | XP_008764146.1 | 7 | 1015 | |||
XM_008765925.1 | XP_008764147.1 | 9 | 912 | |||
XM_008765926.1 | XP_008764148.1 | 8 | 1248 | |||
GenBank mRNA | BC168965.1 | - | 7 | 825 |
