siRNA Details
Gene Symbol: Olr1139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000427.1 | NP_001000427.1 | 1 | 784 |
Gene Symbol: LOC100911988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766029.1 | XP_008764251.1 | 1 | 784 |
